Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody detects endogenous levels of total SYNPR / Synaptoporin protein.
Synaptophysin 2, also known as synaptoporin, is a component of the synaptic vesicle membrane and thought to play an important role in synaptic vesicle trafficking (Dai et al., 2003). It is highly homologous to synaptophysin 1, yet differs in expression. Whereas synaptophysin 1 is ubiquitously expressed, synaptophysin 2 is concentrated in the mossy fiber synapses of the hippocampus (Grabs et al., 1994; Singec et al., 2002).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Synaptoporin; synaptorin
Gene Aliases: 1500003F20Rik; SPO; SYNPR
UniProt ID: (Human) Q8TBG9, (Rat) P22831, (Mouse) Q8BGN8
Entrez Gene ID: (Human) 132204, (Rat) 66030, (Mouse) 72003
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support