Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Positive Samples: HeLa, HepG2, C6, Mouse kidney
Immunogen sequence: MASGRRAPRT GLLELRAGAG SGAGGERWQR VLLSLAEDVL TVSPADGDPG PEPGAPREQE PAQLNGAAEP GAGPPQLPEA LLLQRRRVTV RKADAGGLGI
Syntrophin alpha-1 (Dystrophin, SNTA1) is a large, rod-like cytoskeletal protein found at the inner surface of muscle fibers. Dystrophin is missing in Duchenne Muscular Dystrophy patients and is present in reduced amounts in Becker Muscular Dystrophy patients. The protein encoded by this gene is a peripheral membrane protein found associated with dystrophin and dystrophin-related proteins. This gene is a member of the syntrophin gene family, which contains at least two other structurally-related genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 59 kDa dystrophin-associated protein A1 acidic component 1; acidic alpha 1 syntrophin; Alpha-1-syntrophin; alpha1-syntrophin; dystrophin-associated protein A1, 59kDa, acidic component; Pro-TGF-alpha cytoplasmic domain-interacting protein 1; RP23-192L3.1; syntrophin, acidic 1; syntrophin, alpha 1 (dystrophin-associated protein A1, 59kDa, acidic component); Syntrophin-1; TACIP1
Gene Aliases: AW228934; dJ1187J4.5; LQT12; SNT1; SNTA1; TACIP1
UniProt ID: (Human) Q13424
Entrez Gene ID: (Human) 6640, (Mouse) 20648, (Rat) 362242
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support