Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSDFSEELKG PVTDDEEVET SVLSGAGMHF PWLQTYVETV AIGGKRRKDF AQTTSACLSF IQEALLKHQW QQAAEYMYSY FQTLEDSDSY KRQAAPEIIW KLGSEILFYH PKSNMESFNT FANRMKNIGV MNYLKISLQH ALYLLHHGML KDAKRNLSEA ETWRHGENTS SREILINLIQ AYKGLLQYYT WSEKKMELSK LDKDDYAYNA VAQDVFNHSW KTSANISALI KIPGVWDPFV KSYVEMLEFY GDRDGAQEVL TNYAYDEKFP SNPNAHIYLY NFLKRQKAPR SKLISVLKIL YQIVPSHKLM LEFHTLLRKS EKEEHRKLGL EVLFGVLDFA GCTKNITAWK YLAKYLKNIL MGNHLAWVQE EWNSRKNWWP GFHFSYFWAK SDWKEDTALA CEKAFVAGLL LGKGCRYFRY ILKQDHQILG KKIKRMKRSV KKYSIVNPRL
Initiation of transcription by RNA polymerase I requires the formation of a complex composed of the TATA-binding protein (TBP) and three TBP-associated factors (TAFs) specific for RNA polymerase I. This complex, also known as SL1, binds to the core promoter of ribosomal RNA genes to position the polymerase properly and acts as a channel for regulatory signals. This gene encodes the smallest SL1-specific TAF. Two transcripts encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RNA polymerase I-specific TBP-associated factor 48 kDa; SL1, 48kD subunit; TAFI48; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kD; TATA box binding protein (TBP)-associated factor, RNA polymerase I, A, 48kDa; TATA box binding protein associated factor 1A; TATA box-binding protein-associated factor 1A; TATA box-binding protein-associated factor RNA polymerase I subunit A; TATA-box binding protein associated factor, RNA polymerase I, A; TBP-associated factor 1A; Transcription factor SL1; Transcription initiation factor SL1/TIF-IB subunit A
Gene Aliases: MGC:17061; mTAFI48; RAFI48; SL1; TAF1A; TAFI48
UniProt ID: (Human) Q15573, (Mouse) P97357
Entrez Gene ID: (Human) 9015, (Mouse) 21339
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support