Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TAX1BP1 Antibodies

          Abnova

          TAX1BP1 Polyclonal Antibody, MaxPab™

          View all (14) TAX1BP1 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TAX1BP1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          TAX1BP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TAX1BP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TAX1BP1 Antibody (H00008887-B01P) in WB

          Western Blot analysis of TAX1BP1 expression in transfected 293T cell line by TAX1BP1 MaxPab polyclonal antibody. Lane 1: TAX1BP1 transfected lysate(86.79 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TAX1BP1 Antibody in Western Blot (WB)

          Product Details

          H00008887-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TAX1BP1 (AAH50358.1, 1 a.a. approximately 789 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MTSFQEVPLQ TSNFAHVIFQ NVAKSYLPNA HLECHYTLTP YIHPHPKDWV GIFKVGWSTA RDYYTFLWSP MPEHYVEGST VNCVLAFQGY YLPNDDGEFY QFCYVTHKGE IRGASTPFQF RASSPVEELL TMEDEGNSDM LVVTTKAGLL ELKIEKTMKE KEELLKLIAV LEKETAQLRE QVGRMERELN HEKERCDQLQ AEQKGLTEVT QSLKMENEEF KKRFSDATSK AHQLEEDIVS VTHKAIEKET ELDSLKDKLK KAQHEREQLE CQLKTEKDEK ELYKVHLKNT EIENTKLMSE VQTLKNIDGN KESVITHFKE EIGRLQLCLA EKENLQRTFL LTTSSKEDTC FLKEQLRKAE EQVQATRQEV VFLAKELSDA VNVRDRTMAD LHTARLENEK VKKQLADAVA ELKLNAMKKD QDKTDTLEHE LRREVEDLKL RLQMAADHYK EKFKECQRLQ KQINKLSDQS ANNNNVFTKK TGNQQKVNDA SVNTDPATSA STVDVKPSPS AAEADFDIVT KGQVCEMTKE IADKTEKYNK CKQLLQDEKA KCNKYADELA KMELKWKEQV KIAENVKLEL AEVQDNYKEL KRSLENPAER KMEGQNSQSP QCFKTCSEQN GYVLTLSNAQ PVLQYGNPYA SQETRDGADG AFYPDEIQRP PVRVPSWGLE DNVVCSQPAR NFSRPDGLED SEDSKEDENV PTAPDPPSQH LRGHGTGFCF DSSFDVHKKC PLCELMFPPN YDQSKFEEHV ESHWKVCPMC SEQFPPDYDQ QVFERHVQTH FDQNVLNFD

          Target Information

          This gene encodes a HTLV-1 tax1 binding protein. The encoded protein interacts with TNFAIP3, and inhibits TNF-induced apoptosis by mediating the TNFAIP3 anti-apoptotic activity. Degradation of this protein by caspase-3-like family proteins is associated with apoptosis induced by TNF. This protein may also have a role in the inhibition of inflammatory signaling pathways. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1-binding protein 1; TRAF6-binding protein

          View more View less

          Gene Aliases: CALCOCO3; PRO0105; T6BP; TAX1BP1; TXBP151

          View more View less

          UniProt ID: (Human) Q86VP1

          View more View less

          Entrez Gene ID: (Human) 8887

          View more View less

          Function(s)
          protein binding kinase binding metal ion binding
          Process(es)
          apoptotic process regulation of tumor necrosis factor-mediated signaling pathway negative regulation of NF-kappaB transcription factor activity negative regulation of type I interferon production negative regulation of apoptotic process
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-ztd5s:80/100.66.134.6:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0