Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
This gene encodes a HTLV-1 tax1 binding protein. The encoded protein interacts with TNFAIP3, and inhibits TNF-induced apoptosis by mediating the TNFAIP3 anti-apoptotic activity. Degradation of this protein by caspase-3-like family proteins is associated with apoptosis induced by TNF. This protein may also have a role in the inhibition of inflammatory signaling pathways. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: liver regeneration-related protein 2; Liver regeneration-related protein LRRG004; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1-binding protein 1; Tax1-binding protein 1 homolog; TRAF6-binding protein; TRAF6-binding protein homolog
Gene Aliases: 1200003J11Rik; 1700069J21Rik; Aa1076; AA930106; CALCOCO3; D6Ertd404e; D6Ertd772e; PRO0105; T6BP; TAX1BP1; TXBP151
UniProt ID: (Human) Q86VP1, (Rat) Q66HA4, (Mouse) Q3UKC1
Entrez Gene ID: (Human) 8887, (Rat) 246244, (Mouse) 52440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support