Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Positive test controls include: OVCAR3, HT-29, U-251MG, 293T, Mouse spleen, Mouse testis, Mouse heart, Mouse kidnay, Rat testis. The target is usually found in the following locations: Mitochondrion.
Immunogen sequence: HPDQEDQFFS LVHEKLGSEL PGLEPALQVD LVWALCVLQQ AREAELQAVL HPEFHIQFLG GKSQKDQNTF QKLLHINATA LLEYPEYSGP LLPASAVAPG PSALDRKVTP LQKELQETLK GLLGSADKGS LEVATQYGWV LDAEVLLDSD GEFLPVRDFV APHLAQPTGS QSPPPGSKRL AFLRWEFPNF NSRSKDLLGR FVLARRHIVA AGFLIVDVPF YEWLELKSEW QKGAYLKDKM RKAVAEELAK
TBRG4 (transforming growth factor beta regulator 4), also known as CPR2 (cell cycle progression restoration protein 2) or FAST 4 (FAST kinase domain-containing protein 4), is a 631 amino acid protein that contains one RAP domain and belongs to the FAST kinase family. TBRG4 is ubiquitously expressed and may have a role in cell cycle progression. Existing as two alternatively spliced isoforms, the gene encoding TBRG4 maps to human chromosome 7p13.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cell cycle progression 2 protein; Cell cycle progression protein 2; Cell cycle progression restoration protein 2; FAST 4; FAST kinase domain-containing protein 4; FAST kinase domains 4; H_TD2522F11.8; Protein TBRG4; transforming growth factor beta regulated gene 4; Transforming growth factor beta regulator 4
Gene Aliases: 2310042P22Rik; AA120735; AA408001; AI527316; CPR2; FASTKD4; KIAA0948; mKIAA0948; R74877; Tb-12; TBRG4
UniProt ID: (Human) Q969Z0, (Mouse) Q91YM4, (Rat) Q5M9G9
Entrez Gene ID: (Human) 9238, (Mouse) 21379, (Rat) 360977
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support