Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MGQEEELLRI AKKLEKMVAR KNTEGALDLL KKLHSCQMSI QLLQTTRIGV AVNGVRKHCS DKEVVSLAKV LIKNWKRLLD SPGPPKGEKG EEREKAKKKE KGLECSDWKP EAGLSPPRKK REDPKTRRDS VDSKSSASSS PKRPSVERSN SSKSKAESPK TPSSPLTPTF ASSMCLLAPC YLTGDSVRDK CVEMLSAALK ADDDYKDYGV NCDKMASEIE DHIYQELKST DMKYRNRVRS RISNLKDPRN PGLRRNVLSG AISAGLIAKM TAEEMASDEL RELRNAMTQE AIREHQMAKT GGTTTDLFQC SKCKKKNCTY NQVQTRSADE PMTTFVLCNE CGNRWKFC
Necessary for efficient RNA polymerase II transcription elongation past template-encoded arresting sites. The arresting sites in DNA have the property of trapping a certain fraction of elongating RNA polymerases that pass through, resulting in locked ternary complexes. Cleavage of the nascent transcript by S-II allows the resumption of elongation from the new 3'-terminus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: rhabdomyosarcoma antigen MU-RMS-40.22; transcription elongation factor A (SII), 3; Transcription elongation factor A protein 3; Transcription elongation factor S-II protein 3; Transcription elongation factor TFIIS.h
Gene Aliases: TCEA3; TFIIS; TFIIS.H; TFIISH
UniProt ID: (Human) O75764
Entrez Gene ID: (Human) 6920
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support