Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: VIAECTLKES AAINQILGRR WHALSREEQA KYYELARKER QLHMQLYPGW SARDNYGKKK RRSREKHQES TTGGKRNAFG TYPEKAAAPA PFLPMTVL
TCF7 (transcriptional factor 7) is a transcriptional activator involved in T-cell lymphocyte differentiation. Necessary for the survival of CD4+ and CD8+ immature thymocytes. Isoforms for TCF7 lacking the N-terminal CTNNB1 binding domain cannot fulfill this role. TCF7 binds to the T-lymphocyte-specific enhancer element (5'-WWCAAAG-3') found in the promoter of the CD3E gene. It may also act as feedback transcriptional repressor of CTNNB1 and TCF7L2 target genes. TLE1, TLE2, TLE3, and TLE4 repress transactivation mediated by TCF7 and CTNNB1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HMG-box; OTTHUMP00000159390; OTTHUMP00000159391; T-cell factor 1; T-cell specific; T-cell-specific transcription factor 1; TCF-7; Transcription factor 7
Gene Aliases: TCF-1; TCF1; TCF7
UniProt ID: (Human) P36402
Entrez Gene ID: (Human) 6932
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support