Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MGEPRAGAAL DDGSGWTGSE EGSEEGTGGS EGAGGDGGPD AEGVWSPDIE QSFQEALAIY PPCGRRKIIL SDEGKMYGRN ELIARYIKLR TGKTRTRKQV SSHIQVLARR KSREIQSKLK DQVSKDKAFQ TMATMSSAQL ISAPSLQAKL GPTGPQASEL FQFWSGGSGP PWNVPDVKPF SQTPFTLSLT PPSTDLPGYE PPQALSPLPP PTPSPPAWQA RGLGTARLQL VEFSAFVEPP DAVDSYQRHL FVHISQHCPS PGAPPLESVD VRQIYDKFPE KKGGLRELYD RGPPHAFFLV KFWADLNWGP SGEEAGAGGS ISSGGFYGVS SQYESLEHMT LTCSSKVCSF GKQVVEKVET ERAQLEDGRF VYRLLRSPMC EYLVNFLHKL RQLPERYMMN SVLENFTILQ VVTNRDTQEL LLCTAYVFEV STSERGAQHH IYRLVRD
TEAD2 is a putative transcription factor that binds to the SPH and GT-IIC enhansons (5'-GTGGAATGT-3'). TEAD2 may be involved in the gene regulation of neural development. TEAD2 binds to the M-CAT motif.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TEA domain family member 2; TEAD-2; Transcriptional enhancer factor TEF-4
Gene Aliases: ETF; TEAD-2; TEAD2; TEF-4; TEF4
UniProt ID: (Human) Q15562
Entrez Gene ID: (Human) 8463
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support