Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen Sequence: MNFNTILEEI LIKRSQQKKK TSPLNYKERL FVLTKSMLTY YEGRAEKKYR KGFIDVSKIK CVEIVKNDDG VIPCQNKYPF QVVHDANTLY IFAPSPQSRD LWVKKLKEEI KNNNNIMIKY HPKFWTDGSY QCCRQTEKLA PGCEKYNLFE SSIRKALPPA PETKKRRPPP PIPLEEEDNS EEIVVAMYDF QAAEGHDLRL ERGQEYLILE KNDVHWWRAR DKYGNEGYIP SNYVTGKKSN NLDQYEWYCR NMNRSKAEQL LRSEDKEGGF MVRDSSQPGL
TEC is a non-receptor protein-tyrosine kinase containing a pleckstrin homology domain and is highly expressed in many hematopoietic cell lines. They are also key players in the regulation of the immune functions. Tec kinase is an integral component of T cell signaling and has a distinct role in T cell activation. This gene may be associated with myelodysplastic syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytoplasmic tyrosine kinase, Dscr28C related; MGC126760; MGC126762; Tyrosine-protein kinase Tec
Gene Aliases: PSCTK4; TEC
UniProt ID: (Human) P42680, (Mouse) P24604
Entrez Gene ID: (Human) 7006, (Rat) 84492, (Mouse) 21682
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support