Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAKLLQPPPK FLPSEWHIAN KNQYHRADAQ RSRSERLVAE SQRLVDEIEK TTRKSQSDVN KKLEQRLEEV QFWKKELDDK LEQLVNVTDD LLIYKIRLEK ALETLKEPLH ITETCLAYRE KRIGIDLVHD TVEHELIKEA EIIQGIMALL TRTLEEASEQ IRMNRSAKYN LEKDLKDKFV ALTIDDICFS LNNNSPNIRY SENAVRIEPN SVSLEDWLDF SSTNVEKADK QRNNSLMLKA LVDRILSQTA NDLRKQCDVV DTAFKNGLKD TKDARDKLAD HLAKVMEEIA SQEKNITALE KAILDQEGPA KVAHTRLETR THRPNVELCR DVAQYRLMKE VQEITHNVAR LKETLAQAQA ELKGLHRRQL ALQEEIQVKE NTIYIDEVLC MQMRKSIPLR DGEDHGVWAG GLRPDAVC
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. This gene is predominantly expressed in the testis and in mouse, tektin 1 mRNA was localized to the spermatocytes and round spermatids in the seminiferous tubules, indicating that it may play a role in spermatogenesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tektin-1
Gene Aliases: TEKT1
UniProt ID: (Human) Q969V4
Entrez Gene ID: (Human) 83659
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support