Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MERVGCTLT TTYAHPRPTP TNFLPAISTM ASSYRDRFPH SNLTHSLSLP WRPSTYYKVA SNSPSVAPYC TRSQRVSENT MLPFVSNRTT FFTRYTPDDW YRSNLTNYQE SNTSRHNSEK LRVDTSRLIQ DKYQQTRKTQ ADTTQNLGER VNDIGFWKSE IIHELDEMIG ETNALTDVKK RLERALMETE APLQVARECL FHREKRMGID LVHDEVEAQL LTEVDTILCC QERMKLHLDK AIAQLAANRA SQHELEKDLS DKQTAYRIND KCHHLRNTSD GVGYFRGVER VDATVSVPES W (1-300 aa encoded by BC031688)
This gene product belongs to the tektin family of proteins. Tektins comprise a family of filament-forming proteins that are coassembled with tubulins to form ciliary and flagellar microtubules. The exact function of this gene is not known. Structural component of ciliary and flagellar microtubules. Forms filamentous polymers in the walls of ciliary and flagellar microtubules. Required for progressive sperm mobility.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tektin-3; testicular microtubules-related protein
Gene Aliases: 4933407G07Rik; TEKT3
UniProt ID: (Human) Q9BXF9, (Mouse) Q6X6Z7, (Rat) Q4V8G8
Entrez Gene ID: (Human) 64518, (Mouse) 71062, (Rat) 287392
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support