Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MEFLGTTQT ASYCGPKKCC GLTSLPAVQA PVIQECYQPY YLPGYRYLNS WRPSLFYKIA NVQTCPDEST STLRPPTILP TLRSALFSRY SPHDWDQSNQ LQVRGAEASR LWASRLTDDS MRLLQDKDQL THQMQEGTCR NLGQRLS (1-146 aa encoded by BC130336)
May be a structural component of the sperm flagellum. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Tektin-5
Gene Aliases: 3300001K11Rik; CT149; TEKT5
UniProt ID: (Human) Q96M29, (Mouse) G5E8A8
Entrez Gene ID: (Human) 146279, (Mouse) 70426
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support