Product References
GRHL3 activates FSCN1 to relax cell-cell adhesions between migrating keratinocytes during wound reepithelialization.
JCI insight
Kashgari G,Venkatesh S,Refuerzo S,Pham B,Bayat A,Klein RH,Ramos R,Ta AP,Plikus MV,Wang PH,Andersen B
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Sep 08 00:00:00 UTC 2021
The IRE1/XBP1 signaling axis promotes skeletal muscle regeneration through a cell non-autonomous mechanism.
eLife
Roy A,Tomaz da Silva M,Bhat R,Bohnert KR,Iwawaki T,Kumar A
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Nov 23 00:00:00 UTC 2021
SnRNA sequencing defines signaling by RBC-derived extracellular vesicles in the murine heart.
Life science alliance
Valkov N,Das A,Tucker NR,Li G,Salvador AM,Chaffin MD,Pereira De Oliveira Junior G,Kur I,Gokulnath P,Ziegler O,Yeri A,Lu S,Khamesra A,Xiao C,Rodosthenous R,Srinivasan S,Toxavidis V,Tigges J,Laurent LC,Momma S,Kitchen R,Ellinor P,Ghiran I,Das S
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Dec 01 00:00:00 UTC 2021
SnRNA sequencing defines signaling by RBC-derived extracellular vesicles in the murine heart.
Life science alliance
Valkov N,Das A,Tucker NR,Li G,Salvador AM,Chaffin MD,Pereira De Oliveira Junior G,Kur I,Gokulnath P,Ziegler O,Yeri A,Lu S,Khamesra A,Xiao C,Rodosthenous R,Srinivasan S,Toxavidis V,Tigges J,Laurent LC,Momma S,Kitchen R,Ellinor P,Ghiran I,Das S
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Dec 01 00:00:00 UTC 2021
Engineered red blood cells carrying PCSK9 inhibitors persistently lower LDL and prevent obesity.
PloS one
Deshycka R,Sudaryo V,Huang NJ,Xie Y,Smeding LY,Choi MK,Ploegh HL,Lodish HF,Pishesha N
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Fri Dec 24 00:00:00 UTC 2021
Hypoxia-sensing CAR T cells provide safety and efficacy in treating solid tumors.
Cell reports. Medicine
Kosti P,Opzoomer JW,Larios-Martinez KI,Henley-Smith R,Scudamore CL,Okesola M,Taher MYM,Davies DM,Muliaditan T,Larcombe-Young D,Woodman N,Gillett CE,Thavaraj S,Maher J,Arnold JN
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Apr 20 00:00:00 UTC 2021
Sex Matters: Physiological Abundance of Immuno-Regulatory CD71+ Erythroid Cells Impair Immunity in Females.
Frontiers in immunology
Mashhouri S,Koleva P,Huynh M,Okoye I,Shahbaz S,Elahi S
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Mon Dec 20 00:00:00 UTC 2021
Lef1 restricts ectopic crypt formation and tumor cell growth in intestinal adenomas.
Science advances
Heino S,Fang S,Lähde M,Högström J,Nassiri S,Campbell A,Flanagan D,Raven A,Hodder M,Nasreddin N,Xue HH,Delorenzi M,Leedham S,Petrova TV,Sansom O,Alitalo K
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Fri Nov 19 00:00:00 UTC 2021
Inhibition of TGFβ improves hematopoietic stem cell niche and ameliorates cancer-related anemia.
Stem cell research & therapy
Wang B,Wang Y,Chen H,Yao S,Lai X,Qiu Y,Cai J,Huang Y,Wei X,Guan Y,Wang T,Wang J,Xiang AP
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Mon Jan 18 00:00:00 UTC 2021
C/EBPα and GATA-2 Mutations Induce Bilineage Acute Erythroid Leukemia through Transformation of a Neomorphic Neutrophil-Erythroid Progenitor.
Cancer cell
Di Genua C,Valletta S,Buono M,Stoilova B,Sweeney C,Rodriguez-Meira A,Grover A,Drissen R,Meng Y,Beveridge R,Aboukhalil Z,Karamitros D,Belderbos ME,Bystrykh L,Thongjuea S,Vyas P,Nerlov C
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Mon May 11 00:00:00 UTC 2020
Deregulated Notch and Wnt signaling activates early-stage myeloid regeneration pathways in leukemia.
The Journal of experimental medicine
Kang YA,Pietras EM,Passegué E
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Mon Mar 02 00:00:00 UTC 2020
A RUNX2 stabilization pathway mediates physiologic and pathologic bone formation.
Nature communications
Kim JM,Yang YS,Park KH,Ge X,Xu R,Li N,Song M,Chun H,Bok S,Charles JF,Filhol-Cochet O,Boldyreff B,Dinter T,Yu PB,Kon N,Gu W,Takarada T,Greenblatt MB,Shim JH
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Fri May 08 00:00:00 UTC 2020
The lysophospholipase D enzyme Gdpd3 is required to maintain chronic myelogenous leukaemia stem cells.
Nature communications
Naka K,Ochiai R,Matsubara E,Kondo C,Yang KM,Hoshii T,Araki M,Araki K,Sotomaru Y,Sasaki K,Mitani K,Kim DW,Ooshima A,Kim SJ
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Thu Sep 17 00:00:00 UTC 2020
Methylation of a CGATA element inhibits binding and regulation by GATA-1.
Nature communications
Yang L,Chen Z,Stout ES,Delerue F,Ittner LM,Wilkins MR,Quinlan KGR,Crossley M
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Fri May 22 00:00:00 UTC 2020
Macrophages fine tune satellite cell fate in dystrophic skeletal muscle of mdx mice.
PLoS genetics
Madaro L,Torcinaro A,De Bardi M,Contino FF,Pelizzola M,Diaferia GR,Imeneo G,Bouchè M,Puri PL,De Santa F
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2019
MyD88 is required for satellite cell-mediated myofiber regeneration in dystrophin-deficient mdx mice.
Human molecular genetics
Gallot YS,Straughn AR,Bohnert KR,Xiong G,Hindi SM,Kumar A
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Mon Oct 01 00:00:00 UTC 2018
Age-specific biological and molecular profiling distinguishes paediatric from adult acute myeloid leukaemias.
Nature communications
Chaudhury S,O'Connor C,Cañete A,Bittencourt-Silvestre J,Sarrou E,Prendergast Á,Choi J,Johnston P,Wells CA,Gibson B,Keeshan K
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Dec 11 00:00:00 UTC 2018
Stem cell factor is selectively secreted by arterial endothelial cells in bone marrow.
Nature communications
Xu C,Gao X,Wei Q,Nakahara F,Zimmerman SE,Mar J,Frenette PS
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Fri Jun 22 00:00:00 UTC 2018
A protocol for generating induced T cells by reprogramming B cells in vivo.
Cell regeneration (London, England)
Weng Q,Hu F,Zhang M,Dong Y,Lv C,Wang Y,Liu X,Wang J
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Sat Sep 01 00:00:00 UTC 2018
Differential cytokine contributions of perivascular haematopoietic stem cell niches.
Nature cell biology
Asada N,Kunisaki Y,Pierce H,Wang Z,Fernandez NF,Birbrair A,Ma'ayan A,Frenette PS
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
GATA4-dependent organ-specific endothelial differentiation controls liver development and embryonic hematopoiesis.
The Journal of clinical investigation
Géraud C,Koch PS,Zierow J,Klapproth K,Busch K,Olsavszky V,Leibing T,Demory A,Ulbrich F,Diett M,Singh S,Sticht C,Breitkopf-Heinlein K,Richter K,Karppinen SM,Pihlajaniemi T,Arnold B,Rodewald HR,Augustin HG,Schledzewski K,Goerdt S
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Mar 01 00:00:00 UTC 2017
GATA Factor-Regulated Samd14 Enhancer Confers Red Blood Cell Regeneration and Survival in Severe Anemia.
Developmental cell
Hewitt KJ,Katsumura KR,Matson DR,Devadas P,Tanimura N,Hebert AS,Coon JJ,Kim JS,Dewey CN,Keles S,Hao S,Paulson RF,Bresnick EH
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Mon Aug 07 00:00:00 UTC 2017
Engineered erythrocytes covalently linked to antigenic peptides can protect against autoimmune disease.
Proceedings of the National Academy of Sciences of the United States of America
Pishesha N,Bilate AM,Wibowo MC,Huang NJ,Li Z,Deshycka R,Bousbaine D,Li H,Patterson HC,Dougan SK,Maruyama T,Lodish HF,Ploegh HL
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Mar 21 00:00:00 UTC 2017
Obesity reversibly depletes the basal cell population and enhances mammary epithelial cell estrogen receptor alpha expression and progenitor activity.
Breast cancer research : BCR
Chamberlin T,D'Amato JV,Arendt LM
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Nov 29 00:00:00 UTC 2017
Integrating Enhancer Mechanisms to Establish a Hierarchical Blood Development Program.
Cell reports
Mehta C,Johnson KD,Gao X,Ong IM,Katsumura KR,McIver SC,Ranheim EA,Bresnick EH
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Sep 19 00:00:00 UTC 2017
Age-dependent modulation of vascular niches for haematopoietic stem cells.
Nature
Kusumbe AP,Ramasamy SK,Itkin T,Mäe MA,Langen UH,Betsholtz C,Lapidot T,Adams RH
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Thu Apr 21 00:00:00 UTC 2016
The LMO2 oncogene regulates DNA replication in hematopoietic cells.
Proceedings of the National Academy of Sciences of the United States of America
Sincennes MC,Humbert M,Grondin B,Lisi V,Veiga DF,Haman A,Cazaux C,Mashtalir N,Affar el B,Verreault A,Hoang T
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Feb 02 00:00:00 UTC 2016
A cell-autonomous tumour suppressor role of RAF1 in hepatocarcinogenesis.
Nature communications
Jeric I,Maurer G,Cavallo AL,Raguz J,Desideri E,Tarkowski B,Parrini M,Fischer I,Zatloukal K,Baccarini M
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Dec 21 00:00:00 UTC 2016
PPAR-α and glucocorticoid receptor synergize to promote erythroid progenitor self-renewal.
Nature
Lee HY,Gao X,Barrasa MI,Li H,Elmes RR,Peters LL,Lodish HF
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Thu Jun 25 00:00:00 UTC 2015
Cis-regulatory mechanisms governing stem and progenitor cell transitions.
Science advances
Johnson KD,Kong G,Gao X,Chang YI,Hewitt KJ,Sanalkumar R,Prathibha R,Ranheim EA,Dewey CN,Zhang J,Bresnick EH
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
Genome-wide association study follow-up identifies cyclin A2 as a regulator of the transition through cytokinesis during terminal erythropoiesis.
American journal of hematology
Ludwig LS,Cho H,Wakabayashi A,Eng JC,Ulirsch JC,Fleming MD,Lodish HF,Sankaran VG
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Fri May 01 00:00:00 UTC 2015
Mitochondrial ATP transporter Ant2 depletion impairs erythropoiesis and B lymphopoiesis.
Cell death and differentiation
Cho J,Seo J,Lim CH,Yang L,Shiratsuchi T,Lee MH,Chowdhury RR,Kasahara H,Kim JS,Oh SP,Lee YJ,Terada N
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Sep 01 00:00:00 UTC 2015
Heme-mediated SPI-C induction promotes monocyte differentiation into iron-recycling macrophages.
Cell
Haldar M,Kohyama M,So AY,Kc W,Wu X,Briseño CG,Satpathy AT,Kretzer NM,Arase H,Rajasekaran NS,Wang L,Egawa T,Igarashi K,Baltimore D,Murphy TL,Murphy KM
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Thu Mar 13 00:00:00 UTC 2014
Chemotherapy-induced bone marrow nerve injury impairs hematopoietic regeneration.
Nature medicine
Lucas D,Scheiermann C,Chow A,Kunisaki Y,Bruns I,Barrick C,Tessarollo L,Frenette PS
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Sat Jun 01 00:00:00 UTC 2013
Selective regulation of lymphopoiesis and leukemogenesis by individual zinc fingers of Ikaros.
Nature immunology
Schjerven H,McLaughlin J,Arenzana TL,Frietze S,Cheng D,Wadsworth SE,Lawson GW,Bensinger SJ,Farnham PJ,Witte ON,Smale ST
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Tue Oct 01 00:00:00 UTC 2013
Concomitant inactivation of Rb and E2f8 in hematopoietic stem cells synergizes to induce severe anemia.
Blood
Hu T,Ghazaryan S,Sy C,Wiedmeyer C,Chang V,Wu L
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Thu May 10 00:00:00 UTC 2012
Gfi1b alters histone methylation at target gene promoters and sites of gamma-satellite containing heterochromatin.
The EMBO journal
Vassen L,Fiolka K,Möröy T
Published figure using TER-119 monoclonal antibody (Product # 46-5921-82) in Flow Cytometry
Wed Jun 07 00:00:00 UTC 2006