Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TETRAN's overexpression in cultured human cells caused resistance to some NSAIDs (Non-steroidal anti-inflammatory drugs), suggesting that TETRAN is an efflux pump for some NSAIDs.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Major facilitator superfamily domain-containing protein 10; Tetracycline transporter-like protein
Gene Aliases: MFSD10; TETRAN; TETTRAN
UniProt ID: (Human) Q14728
Entrez Gene ID: (Human) 10227
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support