Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: activating enhancer binding protein 2 beta-like 1; activating enhancer-binding protein 2 delta; Activating enhancer-binding protein 2-delta; AP-2 like; AP2-delta; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); Transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta
Gene Aliases: Tcfap2d; TFAP2BL1; TFAP2D
UniProt ID: (Human) Q7Z6R9, (Mouse) Q91ZK0
Entrez Gene ID: (Human) 83741, (Rat) 301284, (Mouse) 226896
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support