Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MPQRPAASNI PVVGSPNPPS THFASQNQHS YSSPPWAGQH NRKGEKNGMG LCRLSMKVWE TVQRKGTTSC QEVVGELVAK FRAASNHASP NESAYDVKNI
This gene encodes a member of the DP family of transcription factors. These factors heterodimerize with E2F proteins to enhance their DNA-binding activity and promote transcription from E2F target genes. This protein functions as a negative regulator and inhibits the DNA binding and transcriptional activities of E2F factors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cancer/testis antigen 30; CT30; E2F-like protein; Hepatocellular carcinoma-associated antigen 661; Transcription factor Dp family member 3; transcription factor Dp family, member 3
Gene Aliases: CT30; DP4; HCA661; TFDP3
UniProt ID: (Human) Q5H9I0
Entrez Gene ID: (Human) 51270
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support