Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MASRIGLRMQ LMREQAQQEE QRERMQQQAV MHYMQQQQQQ QQQQLGGPPT PAINTPVHFQ SPPPVPGEVL KVQSYLENPT SYHLQQSQHQ KVREYLSETY GNKFAAHISP AQGSPKPPPA ASPGVRAGHV LSSSAGNSAP NSPMAMLHIG SNPERELDDV IDNIMRLDDV LGYINPEMQM PNTLPLSSSH LNVYSSDPQV TASLVGVTSS SCPADLTQKR ELTDAESRAL AKERQKKDNH NLIERRRRFN INDRIKELGM LIPKANDLDV RWNKGTILKA SVDYIRRMQK DLQKSRELEN HSRRLEMTNK QLWLRIQELE MQARVHGLPT TSPSGMNMAE LAQQVVKQEL PSEEGPGEAL MLGAEVPDPE PLPALPPQAP LPLPTQPPSP FHHLDFSHSL SFGGREDEGP PGYPEPLAPG HGSPFPSLSK KDLDLMLLDD SLLPLASDPL LSTMSPEASK ASSRRSSFSM EEGDVL
The Transcription factor EB (TFEB) is a member in the basic helix-loop-helix leucine zipper superfamily of transcription factors that is translocated in a subset of renal tumors. Recent studies have shown that lysosomal biogenesis is regulated by TFEB, which is in turn regulated by the mammalian target of rapamycin (mTOR) complex 1. Other evidence suggests that TFEB coordinates the major steps of the autophagic pathway by driving the expression of autophagy and lysosomal genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha TCFEB; bHLHe35; Class E basic helix-loop-helix protein 35; RP4-696P19.3; T-cell transcription factor EB; Transcription factor EB
Gene Aliases: ALPHATFEB; BHLHE35; TCFEB; TFEB
UniProt ID: (Human) P19484
Entrez Gene ID: (Human) 7942
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support