Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The NELF complex of proteins interacts with the DSIF protein complex to repress transcriptional elongation by RNA polymerase II. The protein encoded by this gene is an essential part of the NELF complex. Alternative translation initiation site usage results in the formation of two isoforms with different N-termini.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Negative elongation factor C/D; negative elongation factor complex member C/D,Th1l; Negative elongation factor D; negative elongation factor proteins C and D; NELF-C/D; NELF-D; TH1 drosophila homolog; TH1 homolog; TH1-like homolog; TH1-like protein; trihydrophobin 1; trihydrophobin 1 homolog
Gene Aliases: 2410003I03Rik; C77797; HSPC130; NELF-C; NELF-D; NELFCD; NELFD; TH1; TH1L
UniProt ID: (Human) Q8IXH7, (Mouse) Q922L6
Entrez Gene ID: (Human) 51497, (Rat) 679203, (Mouse) 57314
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support