Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Antibody detects endogenous levels of total MED24.
This gene encodes a component of the mediator complex, a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator-recruited cofactor 100 kDa component; ARC100; Cofactor required for Sp1 transcriptional activation subunit 4; cofactor required for Sp1 transcriptional activation, subunit 4, 100kDa; CRSP complex subunit 4; DRIP100; genetic suppressor element 2; hTRAP100; Mediator complex subunit 24; Mediator of RNA polymerase II transcription subunit 24; mediator of RNA polymerase II transcription, subunit 24 homolog; MGC8748; mTRAP100; peroxisome proliferation activation receptor binding protein 2; thyroid hormone receptor associated protein 4; thyroid hormone receptor-associated protein 100 kDa; Thyroid hormone receptor-associated protein 4; Thyroid hormone receptor-associated protein complex 100 kDa component; Trap100; Vitamin D3 receptor-interacting protein complex 100 kDa component; vitamin D3 receptor-interacting protein complex component DRIP100
Gene Aliases: 911GSE; ARC100; AU040102; AW547152; CRSP100; CRSP4; D11Ertd307e; DRIP100; Gse2; KIAA0130; MED24; MED5; Pparb2; Pparbp2; R75526; THRAP4; TRAP100
UniProt ID: (Human) O75448, (Rat) Q4V8B3, (Mouse) Q99K74
Entrez Gene ID: (Human) 9862, (Rat) 619436, (Mouse) 23989
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support