Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TIAM1 Antibodies

          Osenses

          TIAM 1 Polyclonal Antibody

          2 References
          View all (12) TIAM1 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TIAM 1 Polyclonal Antibody

          Product Details

          OST00086G-500UG

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (IHC)

          10-50 µg/mL
          -

          Miscellaneous PubMed (MISC)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide from human T-lymphoma invasion and metastasis-inducing protein 1 (TIAM-1, TIAM1) conjugated to an immunogenic carrier protein was used as the antigen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Purification

          Ammonium sulfate precipitation

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 500 µL of sterile water. Centrifuge to remove any insoluble material.

          Specificity of this antibody: TIAM1.

          Target Information

          Modulates the activity of RHO-like proteins and connects extracellular signals to cytoskeletal activities. Acts as a GDP-dissociation stimulator protein that stimulates the GDP-GTP exchange activity of RHO-like GTPases and activates them. Activates RAC1, CDC42, and to a lesser extent RHOA. Required for normal cell adhesion and cell migration.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: human T-lymphoma invasion and metastasis inducing TIAM1 protein; Rho guanine nucleotide exchange factor TIAM1; T-cell lymphoma invasion and metastasis 1; T-lymphoma invasion and metastasis-inducing protein 1; TIAM-1

          View more View less

          Gene Aliases: AI847750; D16Ium10; D16Ium10e; TIAM1

          View more View less

          UniProt ID: (Human) Q13009

          View more View less

          Entrez Gene ID: (Human) 7074, (Mouse) 21844

          View more View less

          Function(s)
          receptor signaling protein activity guanyl-nucleotide exchange factor activity Rho guanyl-nucleotide exchange factor activity protein binding microtubule binding lipid binding kinase binding Rac guanyl-nucleotide exchange factor activity receptor tyrosine kinase binding Rac GTPase binding phospholipid binding ephrin receptor binding
          Process(es)
          cardiac muscle hypertrophy apoptotic process cell-matrix adhesion cell migration Rac protein signal transduction positive regulation of protein binding regulation of Rho protein signal transduction response to cocaine positive regulation of apoptotic process positive regulation of JUN kinase activity ephrin receptor signaling pathway positive regulation of axonogenesis regulation of small GTPase mediated signal transduction Wnt signaling pathway, planar cell polarity pathway positive regulation of dendritic spine morphogenesis regulation of insulin secretion involved in cellular response to glucose stimulus regulation of ERK1 and ERK2 cascade protein localization to membrane activation of GTPase activity positive regulation of Schwann cell chemotaxis regulation of dopaminergic neuron differentiation neuron projection extension regulation of non-canonical Wnt signaling pathway signal transduction small GTPase mediated signal transduction positive regulation of neuron projection development brain-derived neurotrophic factor receptor signaling pathway intracellular signal transduction regulation of GTPase activity positive regulation of GTPase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0