Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAAAALRSGW CRCPRRCLGS GIQFLSSHNL PHGSTYQMRR PGGELPLSKS YSSGNRKGFL SGLLDNVKQE LAKNKEMKES IKKFRDEARR LEESDVLQEA RRKYKTIESE TVRTSEVLRK KLGELTGTVK ESLHEVSKSD LGRKIKEGVE EAAKTAKQSA ESVSKGGEKL GRTAAFRALS QGVESVKKEI DDSVLGQTGP YRRPQRLRKR TEFAGDKFKE EKVFEPNEEA LGVVLHKDSK WYQQWKDFKE NNVVFNRFFE MKMKYDESDN AFIRASRALT DKVTDLLGGL FSKTEMSEVL TEILRVDPAF DKDRFLKQCE NDIIPNVLEA MISGELDILK DWCYEATYSQ LAHPIQQAKA LGLQFHSRIL DIDNVDLAMG KMMEQGPVLI ITFQAQLVMV VRNPKGEVVE GDPDKVLRML YVWALCRDQD ELNPYAAWRL LDISASSTEQ IL
This gene encodes a peripheral membrane protein associated with the mitochondrial inner membrane translocase, which functions in the import of proteins across the mitochondrial inner membrane and into the mitochondrial matrix. The encoded protein mediates binding of mitochondrial heat shock protein 70 to the translocase of inner mitochondrial membrane 23 (TIM23) complex. Expression of this gene is upregulated in kidney in a mouse model of diabetes. A mutation in this gene is associated with familial oncocytic thyroid carcinoma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import inner membrane translocase subunit TIM44; translocase of inner mitochondrial membrane 44 homolog
Gene Aliases: MIMT44; TIM44; TIMM44
UniProt ID: (Human) O43615
Entrez Gene ID: (Human) 10469
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support