Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TIP47 Antibodies

          Progen

          TIP47/PLIN3 Monoclonal Antibody (49.19)

          View all (26) TIP47 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TIP47/PLIN3 Monoclonal Antibody (49.19)

          • Antibody Testing Data (11)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TIP47/PLIN3 Antibody (651168PROGEN) in IHC

          Immunohistochemistry of human colon (courtesy of J.H, University Hospital Heidelberg). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Immunohistochemistry (IHC)
          TIP47/PLIN3 Antibody in Western Blot (WB)

          Product Details

          651168PROGEN

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:100
          -

          Immunohistochemistry (IHC)

          Assay-dependent
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:10
          -
          Product Specifications

          Species Reactivity

          Bovine, Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          49.19

          Immunogen

          Recombinant Polypeptide Of Human Tip47 / Pp17 / Plin3
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          Conc. Not Determined

          Contains

          0.09% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          TIP47 (tail-interacting protein of 47 kD; also named Perilipin 3, PLIN3) is involved in lipid droplet maturation (see e.g. Bulankina 2003 and 2009). The protein has been localized in milk fat globule membranes of human and bovine origin. It has been described also as a placental protein. Increased amounts of TIP47/PLIN3 are secreted into circulation of cervix carcinoma patients. TIP47/PLIN3 shows about 40% sequence homology to adipophilin (PLIN2). Specific for TIP47/PLIN3 and does not cross-react with adipophilin (PLIN2) and perilipin (PLIN1) or other proteins of the PLIN/PAT family. Reactivity on cultured cell lines: PLC, A431, HT29, HeLa, SV-80; human pre-adipocytes.

          Target Information

          TIP47 has been described as cargo protein involved in the trafficking of the mannose-6-phosphate receptor between endosomes and the Golgi complex. Mannose 6-phophate receptors (MPRs) deliver lysosomal hydrolase from the Golgi to endosomes and then return to the Golgi complex. The protein encoded by this gene interacts with the cytoplasmic domains of both cation-independent and cation-dependent MPRs and is required for endosome to Golgi transport. This protein also binds directly to the GTPase RAB9, a member of the RAS oncogene family. The interaction with RAB9 has been shown to increase the affinity of this protein for its cargo. TIP47 has been localized in milk fat globule membranes of human and bovine origin. It has been described as a placental protein, as well. Increased amounts of TIP47 are secreted into circulation of cervix carcinoma patients. Therefore, this protein is probably significant as an oncodevelopmental marker.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 47 kDa mannose 6-phosphate receptor-binding protein; 47 kDa MPR-binding protein; Cargo selection protein TIP47; mannose 6 phosphate receptor binding protein 1; mannose-6-phosphate receptor binding protein 1; Mannose-6-phosphate receptor-binding protein 1; MGC11117; MGC2012; perilipin 3; Perilipin-3; Placental protein 17; PP17; tail-interacting protein, 47 kD; testicular tissue protein Li 114

          View more View less

          Gene Aliases: 1300012C15Rik; BOS_7710; M6PRBP1; PLIN3; PP17; TIP47

          View more View less

          UniProt ID: (Human) O60664, (Mouse) Q9DBG5

          View more View less

          Entrez Gene ID: (Human) 10226, (Bovine) 767984, (Mouse) 66905, (Rat) 316130

          View more View less

          Function(s)
          protein binding cadherin binding involved in cell-cell adhesion
          Process(es)
          vesicle-mediated transport cell-cell adhesion transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0