Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: DPPPHMRVP TIPPNLAGIP GGKPAYSFHV TADGQMQPVP FPPDALIGPG IPRHARQINT LNHGEVVCAV TISNPTRHVY TGGKGCVKVW DISHPGNKSP VSQLDCLNRD NYIRSCKLLP DGCTLIVGGE ASTLSIWDLA APTPRIKAEL TSSAPACYAL AISPDSKVCF SCCSDGNIAV WDLHNQTLVR QFQGHTDGAS CIDISNDGTK LWTGGLDNTV RSWDLREGRQ LQQHDFTSQI FSLGYCPTGE WLAVGMESSN VEVLHVNKPD KYQLHLHESC VLSLKFAYCG KWFVSTGKDN LLNAWRTPYG ASIFQSKESS SVLSCDISVD DKYIVTGSGD KKATVYEVIY (422-770 aa encoded by BC010100)
Transcriptional corepressor that binds to a number of transcription factors. Inhibits NF-kappa-B-regulated gene expression. Inhibits the transcriptional activation mediated by FOXA2, and by CTNNB1 and TCF family members in Wnt signaling. The effects of full-length TLE family members may be modulated by association with dominant-negative AES. Unusual function as coactivator for ESRRG.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E(Sp1) homolog; Enhancer of split groucho-like protein 1; ESG1; RP11-203D19.1; TLE 1; transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); Transducin-like enhancer protein 1
Gene Aliases: ESG; ESG1; GRG1; TLE1
UniProt ID: (Human) Q04724
Entrez Gene ID: (Human) 7088
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support