Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MPHTLWMVWV LGVIISLSKE ESSNQASLSC DRNGICKGSS GSLNSIPSGL TEAVKSLDLS NNRITYISNS DLQRCVNLQA LVLTSNGINT IEEDSFSSLG SLEHLDLSYN YLSNLSSSWF KPLSSLTFLN LLGNPYKTLG ETSLFSHLTK LQILRVGNMD TFTKIQRKDF AGLTFLEELE IDASDLQSYE PKSLKSIQNV SHLILHMKQH ILLLEIFVDV TSSVECLELR DTDLDTFHFS ELSTGETNSL IKKFTFRNVK ITDESLFQVM KLLNQISGLL ELEFDDCTLN GVGNFRASDN DRVIDPGKVE TLTIRRLHIP RFYLFYDLST LYSLTERVKR ITVENSKVFL VPCLLSQHLK SLEYLDLSEN LMVEEYLKNS ACEDAWPSLQ TLILRQNHLA SLEKTGETLL TLKNLTNIDI SKNSFHSMPE TCQWPEKMKY LNLSSTRIHS VTGCIPKTLE ILDVSNNNLN LFSLNLPQLK ELYISRNKLM TLPDASLLPM LLVLKISRNA ITTFSKEQLD SFHTLKTLEA GGNNFICSCE FLSFTQEQQA LAKVLIDWPA NYLCDSPSHV RGQQVQDVRL SVSECHRTAL VSGMCCALFL LILLTGVLCH RFHGLWYMKM MWAWLQAKRK PRKAPSRNIC YDAFVSYSER DAYWVENLMV QELENFNPPF KLCLHKRDFI PGKWIIDNII DSIEKSHKTV FVLSENFVKS EWCKYELDFS HFRLFDENND AAILILLEPI EKKAIPQRFC KLRKIMNTKT YLEWPMDEAQ REGFWVNLRA AIKS
TLR2 is a member of the Toll-like receptor (TLR) family which play a fundamental role in pathogen recognition and activation of innate immunity. TLR proteins act through adaptor molecules such as MyD88 and TIRAP to activate various kinases and transcription factors such as Protein Kinase C (PKC) alpha/beta and NF-kappa-B. TLR2 can form heterodimers with either TLR1 or TLR6, and as a heterodimer, can recognize a variety of bacterial and mycoplasma lipoproteins respectively. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. The various TLRs exhibit different patterns of expression. TLR2 is expressed most abundantly in peripheral blood leukocytes, and mediates host response to Gram-positive bacteria and yeast via stimulation of NF-kappaB. TLR2 aids in the recognition of pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. Ten human homologs of TLRs (TLR1-10) have been described. TLR2 is expressed in human cells such as tonsils, microglia, lymph nodes, and appendices, activated B-cells in germinal centers. CD14+ monocytes express the highest level of TLR2 followed by CD15+ granulocytes, CD19+ B-cells, and CD3+ T-cells. The expression of TLR2 on different cell types are regulated by different immune response modifiers. For example, LPS, GM-CSF, IL-1, and IL-10 up regulates TLR2 whereas IL-4, IFN-gamma, and TNF down regulate TLR2 expression in monocytes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD282; TLR-2; Toll-like receptor 2; Toll/interleukin-1 receptor-like protein 4
Gene Aliases: CD282; TIL4; TLR2
UniProt ID: (Human) O60603
Entrez Gene ID: (Human) 7097
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support