Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of total TM2D1.
Proteolytic cleavage of the Amyloid protein precursor (APP) gives rise to the β-Amyloid and Amyloid A4 proteins, which are present in human platelets. Amyloid deposition is associated with type II diabetes, Down syndrome and a variety of neurological disorders, including Alzheimer's disease. Proteolytic cleavage of APP leads to the formation of the Amyloid β/A4 Amyloid protein. This protein is involved in the formation of neurofibrillary tangles and plaques that characterize the senile plaques of Alzheimer's patients. BBP (Beta-amyloid-binding protein), also known as TM2D1 (TM2 domain-containing protein 1), is a 207 amino acid multi-pass membrane protein containing a G protein-coupling module that allows for interaction with the β-amyloid peptide of APP. In cell culture, expression of BBP induces caspase-dependent vulnerability to β-amyloid peptide toxicity, suggesting that it is a target of β-amyloid and may be involved in the molecular pathophysiology of Alzheimer's disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Amyloid-beta-binding protein; beta-amyloid binding protein; beta-amyloid-binding protein; hBBP; mBBP; TM2 domain-containing protein 1
Gene Aliases: 2310026L18Rik; AA990549; BBP; TM2D1
UniProt ID: (Human) Q9BX74, (Mouse) Q99MB3
Entrez Gene ID: (Human) 83941, (Mouse) 94043, (Rat) 362545
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support