Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MKRCRSDEL QQQQGEEDGA GLEDAASHLP GADLRPGETT GANSAGGPTS DAGAAAAPNP GPRSKPPDLK KIQQLSEGSM FGHGLKHLFH SRRRSREREH QTSQDSQQHQ QQQGMSDHDS PDEKERSPEM HRVSYAMSLH DLPARPTAFN RVLQQIRSRP SIKRGASLHS SSGGGSSGSS SRRTKSSSLE PQRGSPHLLR KAPQDSSLAA ILHQHQCRPR SSSTTDTALL LADG (1-233 aa encoded by BC131629)
TMCC2 may be involved in the regulation of the proteolytic processing of the amyloid precursor protein (APP) possibly also implicating APOE.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cerebral protein 11; transmembrane and coiled-coil domains 2; Transmembrane and coiled-coil domains protein 2
Gene Aliases: 1110063G11Rik; hucep-11; HUCEP11; KIAA0481; RGD1311960; TMCC2
UniProt ID: (Human) O75069, (Mouse) Q80W04
Entrez Gene ID: (Human) 9911, (Rat) 305095, (Mouse) 68875
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support