Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The function of this protein remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Gene signature 188; hepatocellular carcinoma-associated antigen 112; kidney expressed gene 2; Kidney-expressed gene 2 protein; Transmembrane protein 176A
Gene Aliases: 0610011I04Rik; AU040201; AU041743; Gs188; Keg2; RGD1310725; Tmem176a
UniProt ID: (Mouse) Q9DCS1, (Rat) Q4G068
Entrez Gene ID: (Mouse) 66058, (Rat) 297077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support