Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thought to stabilize the calcium channel in an inactivated (closed) state. Modulates calcium current when coexpressed with CACNA1G.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neuronal voltage-gated calcium channel gamma-like subunit; TMEM37; Transmembrane protein 37; voltage-dependent calcium channel gamma subunit-like protein; Voltage-dependent calcium channel gamma-like subunit
Gene Aliases: PR; PR1; TMEM37
UniProt ID: (Human) Q8WXS4
Entrez Gene ID: (Human) 140738
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support