Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
A suggested positive control is Raji cell lysate.
PA5-20550 can be used with blocking peptide PEP-0670.
TNPO3 is a member of the importin beta/transportin family of nuclear import receptors and is specific for the essential splicing factors termed SR proteins, binding specifically and directly to the arginine/serine (RS)-rich domains of these proteins. Further experiments demonstrated that this interaction occurs only when these RS domains are phosphorylated. More recently, TNPO3 was identified as an HIV dependency factor (HDF) and that depletion of TNPO3 resulted in HIV inhibition after reverse transcription but before integration, suggesting that TNPO3 may be an important drug target in HIV treatment. At least three isoforms of TNPO3 are known to exist.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Imp12; importin 12; Importin-12; limb girdle muscular dystrophy 1F (autosomal dominant); Transportin-3; Transportin-SR; TRN-SR
Gene Aliases: 5730544L10Rik; C430013M08Rik; C81142; D6Ertd313e; IPO12; LGMD1F; mKIAA4133; MTR10A; TNPO3; TRN-SR; TRN-SR2; TRNSR
UniProt ID: (Human) Q9Y5L0, (Mouse) Q6P2B1
Entrez Gene ID: (Human) 23534, (Rat) 296954, (Mouse) 320938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support