Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Sequence of this protein is as follows: MQLEIQVALN FIISYLYNKL PRRRVNIFGE ELERLLKKKY EGHWYPEKPY KGSGFRCIHI GEKVDPVIEQ ASKESGLDID DVRGNLPQDL SVWIDPFEVS YQIGEKGPVK VLYVDDNNEN GCELDKEIKN SFNPEAQVFM PISDPASSVS SSPSPPFGHS AAVSPTFMPR STQPLTFTTA TFAATKFGST KMKNSGRSNK VARTSPINLG LNVNDLLKQK AISSSMHSLY GLGLGSQQQP QQQQQPAQPP PPPPPPQQQQ QQKTSALSPN AKEFIFPNMQ GQGSSTNGMF PGDSPLNLSP LQYSNAFDVF AAYGGLNEKS FVDGLNFSLN NMQYSNQQFQ PVMAN
This gene encodes a member of the tob/btg1 family of anti-proliferative proteins that have the potential to regulate cell growth. When exogenously expressed, this protein supresses cell growth in tissue culture. The protein undergoes phophorylation by a serine/threonine kinase, 90 kDa ribosomal S6 kinase. Interactions of this protein with the v-erb-b2 erythroblastic leukemia viral oncogene homolog 2 gene product p185 interferes with growth suppression. This protein inhibits T cell proliferation and transcription of cytokines and cyclins. The protein interacts with both mothers against decapentaplegic Drosophila homolog 2 and 4 to enhance their DNA binding activity. This interaction inhibits interleukin 2 transcription in T cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC104792; MGC34446; proliferation-inducing gene 49; Protein Tob1; Transducer of erbB-2 1
Gene Aliases: APRO5; APRO6; PIG49; TOB; TOB1; TROB; TROB1
UniProt ID: (Human) P50616
Entrez Gene ID: (Human) 10140
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support