Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAFGKSHRDP YATSVGHLIE KATFAGVQTE DWGQFMHICD IINTTQDGPK DAVKALKKRI SKNYNHKEIQ LTLSLIDMCV QNCGPSFQSL IVKKEFVKEN LVKLLNPRYN LPLDIQNRIL NFIKTWSQGF PGGVDVSEVK EVYLDLVKKG VQFPPSEAEA ETARQETAQI SSNPPTSVPT APALSSVIAP KNSTVTLVPE QIGKLHSELD MVKMNVRVMS AILMENTPGS ENHEDIELLQ KLYKTGREMQ ERIMDLLVVV ENEDVTVELI QVNEDLNNAI LGYERFTRNQ QRILEQNKNQ KEATNTTSEP SAPSQDLLDL SPSPRMPRAT LGELNTMNNQ LSGLSK
TOM1L1 is an adapter protein involved in signaling pathways. Interacts with the SH2 and SH3 domains of various signaling proteins when it is phosphorylated. May promotes FYN activation, possibly by disrupting intramolecular SH3-dependent interactions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OK/KNS-CL.3; Src-activating and signaling molecule protein; Target of Myb-like protein 1; target of myb1 (chicken)-like 1; TOM1-like protein 1
Gene Aliases: SRCASM; TOM1L1
UniProt ID: (Human) O75674
Entrez Gene ID: (Human) 10040
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support