Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TOP3B Antibodies

          Abnova

          TOP3B Monoclonal Antibody (1H4)

          View all (9) TOP3B antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TOP3B Monoclonal Antibody (1H4)

          • Antibody Testing Data (1)
          TOP3B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TOP3B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TOP3B Antibody (H00008940-M04) in WB

          Western Blot analysis of TOP3B expression in transfected 293T cell line by TOP3B monoclonal antibody (M04), clone 1H4. Lane 1: TOP3B transfected lysate(96.662 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TOP3B Antibody in Western Blot (WB)

          Product Details

          H00008940-M04

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -

          ELISA (ELISA)

          10 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG2a, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1H4

          Immunogen

          TOP3B (AAH02432, 1 a.a. approximately 862 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          Immunogen sequence: MKTVLMVAEK PSLAQSIAKI LSRGSLSSHK GLNGACSVHE YTGTFAGQPV RFKMTSVCGH VMTLDFLGKY NKWDKVDPAE LFSQAPTEKK EANPKLNMVK FLQVEGRGCD YIVLWLDCDK EGENICFEVL DAVLPVMNKA HGGEKTVFRA RFSSITDTDI CNAMACLGEP DHNEALSVDA RQELDLRIGC AFTRFQTKYF QGKYGDLDSS LISFGPCQTP TLGFCVERHD KIQSFKPETY WVLQAKVNTD KDRSLLLDWD RVRVFDREIA QMFLNMTKLE KEAQVEATSR KEKAKQRPLA LNTVEMLRVA SSSLGMGPQH AMQTAERLYT QGYISYPRTE TTHYPENFDL KGSLRQQANH PYWANTVKRL LAEGINRPRK GHDAGDHPPI TPMKSATEAE LGGDAWRLYE YITRHFIATV SHDCKYLQST ISFRIGPELF TCSGKTVLSP GFTEVMPWQS VPLEESLPTC QRGDAFPVGE VKMLEKQTNP PDYLTEAELI TLMEKHGIGT DASIPVHINN ICQRNYVTVE SGRRLKPTNL GIVLVHGYYK IDAELVLPTI RSAVEKQLNL IAQGKADYRQ VLGHTLDVFK RKFHYFVDSI AGMDELMEVS FSPLAATGKP LSRCGKCHRF MKYIQAKPSR LHCSHCDETY TLPQNGTIKL YKELRCPLDD FELVLWSSGS RGKSYPLCPY CYNHPPFRDM KKGMGCNECT HPSCQHSLSM LGIGQCVECE SGVLVLDPTS GPKWKVACNK CNVVAHCFEN AHRVRVSADT CSVCEAALLD VDFNKAKSPL PGDETQHMGC VFCDPVFQEL VELKHAASCH PMHRGGPGRR QGRGRGRARR PPGKPNPRRP KDKMSALAAY FV

          Target Information

          DNA topoisomerases are nuclear enzymes that regulate the topological structure of DNA by transiently breaking and rejoining DNA strands. Although DNA topoisomerase I and DNA topoisomerase II relax both positive and negative supercoils, DNA topoisomerase III relaxes only negative supercoils. DNA topoisomerase III alpha exists as a long and a short isoform, which are produced by alternative splicing. DNA topoisomerase III alpha, which localizes to the nucleolus, is constitutively expressed and remains at high levels throughout the cell cycle in HL-60 cells. DNA topoisomerase III beta exists as three isoforms, namely beta1, beta2 and beta3, also produced by alternative splicing. DNA topoisomerase III beta1 is expressed in testes, heart and skeletal muscle, whereas beta2 is expressed in thymus, kidney and pancreas.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DNA topoisomerase 3-beta-1; DNA topoisomerase III beta-1; topoisomerase III beta

          View more View less

          Gene Aliases: TOP3B; TOP3B1

          View more View less

          UniProt ID: (Human) O95985

          View more View less

          Entrez Gene ID: (Human) 8940

          View more View less

          Function(s)
          DNA binding DNA topoisomerase activity DNA topoisomerase type I activity protein binding poly(A) RNA binding
          Process(es)
          DNA topological change chromosome segregation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0