Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
Immunogen Sequence: MDRGQPSLEP AAAAPRASGR CVIAPVRAVL RLRRRVCVLR KRRLLQPGGG PDVGTGAPRP GCSPRAPRAD LDQPKFFTFD SPAELPSRTP RKKRRRSRLV LYPETSRKYR PRVEHRSRAQ RCLLLLVAIV
TOR4A gene ontology annotations related to this gene include platelet degranulation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Torsin family 4 member A; torsin family 4, member A; torsin family protein C9orf167; torsin family protein C9orf167 homolog; Torsin-4A
Gene Aliases: A830007P12Rik; C9orf167; TOR4A
UniProt ID: (Human) Q9NXH8, (Mouse) Q8BH02
Entrez Gene ID: (Human) 54863, (Mouse) 227612
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support