Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: SETLSQAGQK ASAAFSSVGS VITKKLEDVK NSPTFKSFEE KVENLKSKVG GTKPAGGDFG EVLNSAANAS ATTTEPLPEK TQESL
It belongs to the TPD52 family. Its exact function remains unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: prostate and colon associated protein; prostate leucine zipper; Protein N8; Tumor protein D52
Gene Aliases: D52; hD52; N8L; PC-1; PrLZ; TPD52
UniProt ID: (Human) D0UFD5
Entrez Gene ID: (Human) 7163
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support