Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MRALAVLSVT LVMACTEAFF PFISRGKELL WGKPEESRVS SVLEESKRLV DTAMYATMQR NLKKRGILSA AQLLSFSKLP EPTSGVIARA AEIMETSIQA MKRKVNLKTQ QSQHPTDALS EDLLSIIANM SGCLPYMLPP KCPNTCLANK YRPITGACNN RDHPRWGASN TALARWLPPV YEDGFSQPRG WNPGFLYNGF PLPPVREVTR HVIQVSNEVV TDDDRYSDLL MAWGQYIDHD IAFTPQSTSK AAFGGGADCQ MTCENQNPCF PIQLPEEARP AAGTACLPFY RSSAACGTGD QGALFGNLST ANPRQQMNGL TSFLDASTVY GSSPALERQL RNWTSAEGLL RVHARLRDSG RAYLPFVPPR APAACAPEPG IPGETRGPCF LAGDGRASEV PSLTALHTLW LREHNRLAAA LKALNAHWSA DAVYQEARKV VGALHQIITL RDYIPRILGP EAFQQYVGPY EGYDSTANPT VSNVFSTAAF RFGHATIHPL VRRLDASFQE HPDLPGLWLH QAFFSPWTLL RGGGLDPLIR GLLARPAKLQ VQDQLMNEEL TERLFVLSNS STLDLASINL QRGRDHGLPG YNEWREFCGL PRLETPADLS TAIASRSVAD KILDLYKHPD NIDVWLGGLA ENFLPRARTG PLFACLIGKQ MKALRDGDWF WWENSHVFTD AQRRELEKHS LSRVICDNTG LTRVPMDAFQ VGKFPEDFES CDSITGMNLE AWRETFPQDD KCGFPESVEN GDFVHCEESG RRVLVYSCRH GYELQGREQL TCTQEGWDFQ PPLCKDVNEC ADGAHPPCHA SARCRNTKGG FQCLCADPYE LGDDGRTCVD SGRLPRVTWI SMSLAALLIG GFAGLTSTVI CRWTRTGTKS TLPISETGGG TPELRCGKHQ AVGTSPQRAA AQDSEQESAG MEGRDTHRLP RAL
Thyroid Peroxidase is a gene that encodes for a membrane-bound glycoprotein, which functions as an enzyme and plays a crucial role in the proper functioning of the thyroid gland. This protein is responsible for the iodination of tyrosine residues in thyroglobulin and the formation of phenoxy-ester between pairs of iodinated tyrosines, which leads to the production of thyroid hormones such as thyroxine and triiodothyronine. Mutations in this gene are known to cause several disorders related to thyroid hormone production, including congenital hypothyroidism, congenital goiter, and thyroid hormone organification defect IIA. Several transcript variants encoding distinct isoforms have been identified for this gene, but the full-length nature of some variants remains undetermined. Diseases associated with TPO include Thyroid Dyshormonogenesis 2A and Congenital Hypothyroidism.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EC 1.11.1; thyroid microsomal antigen; Thyroid peroxidase; thyroperoxidase; TPO; TPXEC 1.11.1.8
Gene Aliases: MSA; TDH2A; TPO; TPX
UniProt ID: (Human) P07202
Entrez Gene ID: (Human) 7173
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support