Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MASEAEKTF HRFAAFGESS SSGTEMNNKN FSKLCKDCGI MDGKTVTSTD VDIVFSKVKA KNARTITFQQ FKEAVKELGQ KRFKGKSPDE VLENIYGLME GKDPATTGAT KATTVGAVDR LTDTSKYTGT HKELFDESGK GKGIAGREEM TDNTGYVSGY KGSGTYDKKT K (1-170 aa encoded by BC038970)
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: p25 beta; Protein p25-beta; TPPP/p18; Tubulin polymerization-promoting protein family member 2
Gene Aliases: C14orf8; CT152; Gm1790; Gm77; P18; p25beta; RGD1566185; TPPP2
UniProt ID: (Human) P59282, (Mouse) Q0P5Y3
Entrez Gene ID: (Human) 122664, (Rat) 498497, (Mouse) 219038
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support