Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TPR Antibodies

          Invitrogen

          TPR Polyclonal Antibody

          View all (14) TPR antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TPR Polyclonal Antibody

          • Antibody Testing Data (7)
          TPR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TPR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TPR Antibody (PA5-54049) in ICC/IF

          Immunofluorecent analysis of TPR in human cell line A-431 using TPR Polyclonal Antibody (Product # PA5-54049). Staining shows localization to nuclear membrane. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TPR Antibody in Immunocytochemistry (ICC/IF)
          TPR Antibody in Immunohistochemistry (IHC)
          TPR Antibody in Immunohistochemistry (IHC)
          TPR Antibody in Immunohistochemistry (IHC)
          TPR Antibody in Immunohistochemistry (IHC)
          TPR Antibody in Immunohistochemistry (IHC)
          TPR Antibody in Western Blot (WB)

          Product Details

          PA5-54049

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.04-0.4 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:50
          -

          Immunocytochemistry (ICC/IF)

          0.25-2 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Recombinant protein corresponding to Human TPR. Recombinant protein control fragment (Product #RP-91827).
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.05 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.2, with 40% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2648856

          Product Specific Information

          Immunogen sequence: RDEEVSSADI SSSSEVISQH LVSYRNIEEL QQQNQRLLVA LRELGETRER EEQETTSSKI TELQLKLESA LTELEQLRKS RQHQMQLVDS IVRQRDMYRI LLSQTTGVAI PLHASSLDDV SLASTPKRPS TSQTVSTPAP

          Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 92%.

          Target Information

          Steroidogenic factor-1 (SF-1) regulates multiple genes involved in the adrenal and gonadal development and in the biosynthesis of a variety of hormones, including adrenal and gonadal steroids, anti-Mullerian hormone (AMH), and gonadotropins. SF-1 belongs to the fushi tarazu factor-1 (FTZ-F1) subfamily of orphan nuclear receptors. In the adult ovary, SF-1 localizes to theca/interstitial cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Megator; NPC-associated intranuclear protein; nuclear pore complex-associated protein TPR; Nucleoprotein TPR; translocated promoter region (to activated MET oncogene); Translocated promoter region protein; tumor potentiating region

          View more View less

          Gene Aliases: TPR

          View more View less

          UniProt ID: (Human) P12270

          View more View less

          Entrez Gene ID: (Human) 7175

          View more View less

          Function(s)
          molecular_function chromatin binding mRNA binding transporter activity nucleocytoplasmic transporter activity protein binding tubulin binding heat shock protein binding protein homodimerization activity protein anchor poly(A) RNA binding mitogen-activated protein kinase binding dynein complex binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter MAPK import into nucleus RNA import into nucleus RNA export from nucleus mRNA export from nucleus tRNA export from nucleus protein import into nucleus protein export from nucleus nuclear pore organization mitotic nuclear division mitotic nuclear envelope disassembly mitotic spindle assembly checkpoint biological_process regulation of mRNA export from nucleus regulation of glucose transport regulation of mitotic sister chromatid separation viral process protein sumoylation viral transcription gene silencing by RNA positive regulation of heterochromatin assembly regulation of protein stability mRNA export from nucleus in response to heat stress cellular response to heat cellular response to interferon-alpha regulation of protein import into nucleus positive regulation of protein import into nucleus nuclear matrix organization negative regulation of translational initiation regulation of protein export from nucleus positive regulation of protein export from nucleus negative regulation of RNA export from nucleus nuclear pore complex assembly cell division response to epidermal growth factor intracellular transport of virus positive regulation of mitotic cell cycle spindle assembly checkpoint positive regulation of intracellular protein transport regulation of cellular response to heat regulation of mitotic spindle assembly
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0