Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: PGCRAQKGI PAALSPGPGP IPAPIPGPAQ IPGPLPGSIP GPIPGPAQIP SPIPAPIPGP ISGPVQIPGP FRGPIPGPIS GPAPIPGPIS GPFSGPNPGP IPGPNPGPIP GPISGPIPGP ISVPIPGPIP GPISGPISGP NPGPIPGPIP GPISGPNPGP IPGPISGPNP GLIPGPIPGP ISGPGPIIGP IPSPAQIPGP GRLQGPGPIL SPGRMRSPGS LPGLAPILGP GSGPGSGSVP APIPGPGSLP APAPLWPQSP DASDFLPDTQ LFPHFTELLL PLDPLEGSSV STMTSQYQEG DDSMGKKHSG SQPQEEGGSV NENHSGPRLL LDL (80-411 aa encoded by BC137501)
Homeobox genes encode DNA-binding proteins, many of which are thought to be involved in early embryonic development. Homeobox genes encode a DNA-binding domain of 60 to 63 amino acids referred to as the homeodomain. This gene is a member of the TPRX homeobox gene family.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CRX like homeobox 2; tetra-peptide repeat homeobox 1; Tetra-peptide repeat homeobox protein 1
Gene Aliases: TPRX; TPRX1
UniProt ID: (Human) Q8N7U7
Entrez Gene ID: (Human) 284355
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support