Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MRLLPRLLLL LLLVFPATVL FRGGPRGLLA VAQDLTEDEE TVEDSIIEDE DDEAEVEEDE PTDLVEDKEE EDVSGEPEAS PSADTTILFV KGEDFPANNI VKFLVGFTNK GTEDFIVESL DASFRYPQDY QFYIQNFTAL PLNTVVPPQR QATFEYSFIP AEPMGGRPFG LVINLNYKDL NGNVFQDAVF NQTVTVIERE DGLDGETIFM YMFLAGLGLL VIVGLHQLLE SRKRKRPIQK VEMGTSSQND VDMSWIPQET LNQINKASPR RLPRKRAQKR SVGSDE
The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781N23103; Signal sequence receptor subunit alpha; signal sequence receptor, alpha; SSR alpha subunit; SSR-alpha; translocon-associated protein alpha subunit; Translocon-associated protein subunit alpha; TRAP alpha; TRAP-alpha
Gene Aliases: PSEC0262; SSR1; TRAPA
UniProt ID: (Human) P43307
Entrez Gene ID: (Human) 6745
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support