Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Predicted to react with bovine based on sequence homology.
The function of C5orf44 has not been widely studied, and is yet to be fully elucidated.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Trafficking protein particle complex subunit 13; Trs65-related; UPF0533 protein C5orf44; UPF0533 protein C5orf44 homolog
Gene Aliases: 2410002O22Rik; 2610524F24Rik; AI451892; AU017229; C5orf44; RGD1306583; TRAPPC13
UniProt ID: (Human) A5PLN9, (Mouse) Q3TIR1, (Rat) Q5M887
Entrez Gene ID: (Human) 80006, (Mouse) 66975, (Rat) 294709
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support