Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASAASVTSL ADEVNCPICQ GTLREPVTID CGHNFCRACL TRYCEIPGPD LEESPTCPLC KEPFRPGSFR PNWQLANVVE NIERLQLVST LGLGEEDVCQ EHGEKIYFFC EDDEMQLCVV CREAGEHATH TMRFLEDAAA PYREQIHKCL KCLRKEREEI QEIQSRENKR MQVLLTQVST KRQQVISEFA HLRKFLEEQQ SILLAQLESQ DGDILRQRDE FDLLVAGEIC RFSALIEELE EKNERPAREL LTDIRSTLIR CETRKCRKPV AVSPELGQRI RDFPQQALPL QREMKMFLEK LCFELDYEPA HISLDPQTSH PKLLLSEDHQ RAQFSYKWQN SPDNPQRFDR ATCVLAHTGI TGGRHTWVWM ARVPGDSGCC QFCSPPSVLG TEVAA
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. This protein localizes to cytoplasmic bodies. Studies in mice suggest that this protein plays a role in terminal differentiation of erythroid cells. Alternate splicing of this gene generates two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B30-RING finger protein; hematopoietic RING finger 1; RING finger protein 9; tripartite motif protein 10; tripartite motif-containing 10; Tripartite motif-containing protein 10; Zn-finger protein
Gene Aliases: HERF1; RFB30; RNF9; TRIM10
UniProt ID: (Human) Q9UDY6
Entrez Gene ID: (Human) 10107
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support