Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MATLVVNKLG AGVDSGRQGS RGTAVVKVLE CGVCEDVFSL QGDKVPRLLL CGHTVCHDCL TRLPLHGRAI RCPFDRQVTD LGDSGVWGLK KNFALLELLE RLQNGPIGQY GAAEESIGIS GESIIRCDED EAHLASVYCT VCATHLCSEC SQVTHSTKTL AKHRRVPLAD KPHEKTMCSQ HQVHAIEFVC LEEGCQTSPL MCCVCKEYGK HQGHKHSVLE PEANQIRASI LDMAHCIRTF TEEISDYSRK LVGIVQHIEG GEQIVEDGIG MAHTEHVPGT AENARSCIRA YFYDLHETLC RQEEMALSVV DAHVREKLIW LRQQQEDMTI LLSEVSAACL HCEKTLQQDD CRVVLAKQEI TRLLETLQKQ QQQFTEVADH IQLDASIPVT FTKDNRVHIG PKMEIRVVTL GLDGAGKTTI LFKLKQDEFM QPIPTIGFNV ETVEYKNLKF TIWDVGGKHK LRPLWKHYYL NTQAVVFVVD SSHRDRISEA HSELAKLLTE KELRDALLLI FANKQDVAGA LSVEEITELL SLHKLCCGRS WYIQGCDARS GMGLYEGLDW LSRQLVAAGV LDVA
Acts as an E3 ubiquitin-protein ligase. In the presence of the human cytomegalovirus (HCMV) protein UL144, participates in 'Lys-63'-linked auto-ubiquitination of TRAF6 resulting in the virally controlled activation of NF-kappa-B at early time of infection. The C-terminus can act as an allosteric activator of the cholera toxin catalytic subunit.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADP-ribosylation factor domain protein 1, 64kDa; ADP-ribosylation factor domain-containing protein 1; ARF domain protein 1; E3 ubiquitin-protein ligase TRIM23; GTP-binding protein ARD-1; RING finger protein 46; RING-type E3 ubiquitin transferase TRIM23; tripartite motif protein TRIM23; tripartite motif-containing 23; Tripartite motif-containing protein 23
Gene Aliases: ARD1; ARFD1; RNF46; TRIM23
UniProt ID: (Human) P36406
Entrez Gene ID: (Human) 373
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support