Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TRIM32 Antibodies

          Abnova

          TRIM32 Polyclonal Antibody, MaxPab™

          View all (10) TRIM32 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite TRIM32 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          TRIM32 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TRIM32 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          TRIM32 Antibody (H00022954-B01P) in WB

          Western Blot analysis of TRIM32 expression in transfected 293T cell line by TRIM32 MaxPab polyclonal antibody. Lane 1: TRIM32 transfected lysate(71.83 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          TRIM32 Antibody in Western Blot (WB)

          Product Details

          H00022954-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          TRIM32 (AAH03154.1, 1 a.a. approximately 653 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAAAASHLN LDALREVLEC PICMESFTEE QLRPKLLHCG HTICRQCLEK LLASSINGVR CPFCSKITRI TSLTQLTDNL TVLKIIDTAG LSEAVGLLMC RSCGRRLPRQ FCRSCGLVLC EPCREADHQP PGHCTLPVKE AAEERRRDFG EKLTRLRELM GELQRRKAAL EGVSKDLQAR YKAVLQEYGH EERRVQDELA RSRKFFTGSL AEVEKSNSQV VEEQSYLLNI AEVQAVSRCD YFLAKIKQAD VALLEETADE EEPELTASLP RELTLQDVEL LKVGHVGPLQ IGQAVKKPRT VNVEDSWAME ATASAASTSV TFREMDMSPE EVVASPRASP AKQRGPEAAS NIQQCLFLKK MGAKGSTPGM FNLPVSLYVT SQGEVLVADR GNYRIQVFTR KGFLKEIRRS PSGIDSFVLS FLGADLPNLT PLSVAMNCQG LIGVTDSYDN SLKVYTLDGH CVACHRSQLS KPWGITALPS GQFVVTDVEG GKLWCFTVDR GSGVVKYSCL CSAVRPKFVT CDAEGTVYFT QGLGLNLENR QNEHHLEGGF SIGSVGPDGQ LGRQISHFFS ENEDFRCIAG MCVDARGDLI VADSSRKEIL HFPKGGGYSV LIREGLTCPV GIALTPKGQL LVLDCWDHCI KIYSYHLRRY STP

          Target Information

          TRIM32 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM32 localizes to cytoplasmic bodies. The protein has also been localized to the nucleus, where it interacts with the activation domain of the HIV-1 Tat protein. The Tat protein activates transcription of HIV-1 genes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 72 kDa Tat-interacting protein; E3 ubiquitin-protein ligase TRIM32; RING-type E3 ubiquitin transferase TRIM32; RP11-67K19.1; TAT-interactive protein, 72-KD; tripartite motif-containing 32; Tripartite motif-containing protein 32; Zinc finger protein HT2A; zinc-finger protein HT2A

          View more View less

          Gene Aliases: BBS11; HT2A; LGMD2H; TATIP; TRIM32

          View more View less

          UniProt ID: (Human) Q13049

          View more View less

          Entrez Gene ID: (Human) 22954

          View more View less

          Function(s)
          transcription coactivator activity RNA binding ubiquitin-protein transferase activity protein binding zinc ion binding ligase activity myosin binding Tat protein binding translation initiation factor binding ubiquitin binding protein self-association ubiquitin protein ligase activity
          Process(es)
          protein polyubiquitination tissue homeostasis actin ubiquitination response to UV protein ubiquitination positive regulation of cell growth positive regulation of cell migration regulation of type I interferon production negative regulation of viral transcription response to tumor necrosis factor protein ubiquitination involved in ubiquitin-dependent protein catabolic process positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response fat cell differentiation positive regulation of neuron differentiation positive regulation of protein catabolic process positive regulation of cell cycle positive regulation of proteolysis muscle cell cellular homeostasis negative regulation of fibroblast proliferation positive regulation of neurogenesis positive regulation of sequence-specific DNA binding transcription factor activity positive regulation of NF-kappaB transcription factor activity positive regulation of striated muscle cell differentiation axon development negative regulation of viral release from host cell negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage positive regulation of tumor necrosis factor-mediated signaling pathway positive regulation of interleukin-17-mediated signaling pathway positive regulation of chemokine (C-C motif) ligand 20 production positive regulation of cell motility
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-bff4f7d6d-tmrcx:80/100.66.135.11:80.
          git-commit: 683fd26f943b2dde6a08880618459b58d20c3896
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-Offline