Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SGFWYVCRTQ GVEGDHCVTS DPATSPLVLA IPRRLRVELE CEEGELSFYD AERHCHLYTF HARFGEVRPY FYLGGARGAG PPEPLRICPL HISVKEELDG
TRIM35 is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. TRIM35 may play a role as a tumor suppressor, and is implicated in the cell death mechanism. The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM35; Hemopoietic lineage switch protein 5; MGC17233; tripartite motif-containing 35
Gene Aliases: HLS5; KIAA1098; MAIR; TRIM35
UniProt ID: (Human) Q9UPQ4
Entrez Gene ID: (Human) 23087
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support