Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TSFEDYVVNT SKQTELLGEL SFFSSGIDVP EINEEQSKVY NNALINWHHP EKDKADSYVL EYRKINRDDE MSWNEIEVCG TSKIIQDLEN SSTYAFRVRA YKGSICSPC
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. Multiple alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM36; RING finger protein 98; RING-type E3 ubiquitin transferase TRIM36; tripartite motif protein 36; tripartite motif-containing 36; Tripartite motif-containing protein 36; Zinc-binding protein Rbcc728
Gene Aliases: HAPRIN; RBCC728; RNF98; TRIM36
UniProt ID: (Human) Q9NQ86
Entrez Gene ID: (Human) 55521
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support