Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASTTSTKKM MEEATCSICL SLMTNPVSIN CGHSYCHLCI TDFFKNPSQK QLRQETFCCP QCRAPFHMDS LRPNKQLGSL IEALKETDQE MSCEEHGEQF HLFCEDEGQL ICWRCERAPQ HKGHTTALVE DVCQGYKEKL QKAVTKLKQL EDRCTEQKLS TAMRITKWKE KVQIQRQKIR SDFKNLQCFL HEEEKSYLWR LEKEEQQTLS RLRDYEAGLG LKSNELKSHI LELEEKCQGS AQKLLQNVND TLSRSWAVKL ETSEAVSLEL HTMCNVSKLY FDVKKMLRSH QVSVTLDPDT AHHELILSED RRQVTRGYTQ ENQDTSSRRF TAFPCVLGCE GFTSGRRYFE VDVGEGTGWD LGVCMENVQR GTGMKQEPQS GFWTLRLCKK KGYVALTSPP TSLHLHEQPL LVGIFLDYEA GVVSFYNGNT GCHIFTFPKA SFSDTLRPYF QVYQYSPLFL PPPGD
This gene encodes a member of the tripartite motif (TRIM) family. The encoded protein contains a RING-type zinc finger, B box-type zinc finger and SPRY domain. The function of this protein has not been identified. A pseudogene of this gene is located on the long arm of chromosome 4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM38; MGC8946; RING finger protein 15; Ro/SSA ribonucleoprotein homolog; Tripartite motif-containing protein 38; Zinc finger protein RoRet
Gene Aliases: RNF15; RORET; TRIM38
UniProt ID: (Human) O00635
Entrez Gene ID: (Human) 10475
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support