Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: RRQQILREF EELHRRLDEE QQVLLSRLEE EEQDILQRLR ENAAHLGDKR RDLAHLAAEV EGKCLQSGFE MLKDVKSTLE KCEKVKTMEV TSVSIELEKN FSNFPRQYFA LRKILKQLIA DVTLDPETAH PNLVLSEDRK SVKFVETRLR DLPDTPRRFT FYPCVLATEG FTSGRHYWEV EVGDKTHWAV GVCRDSVSRK GELTPLPETG YWRVRLWNGD KYAATTTPFT PLHIKVKPKR VGIFLDYEAG TLSFYNVTDR SHIYTFTDTF TEKLWPLFYP GIRAGRKNAA PLTIRPPTDW E (189-489 aa encoded by BC034985)
The protein encoded by this gene is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The function of this protein has not been identified. This gene lies within the major histocompatibility complex class I region on chromosome 6. Alternate splicing results in two transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM39; RING finger protein 23; RING-B box-coiled coil-B30.2; RING-B box-coiled-coil-B30.2; RING-type E3 ubiquitin transferase TRIM39; Testis-abundant finger protein; tripartite motif protein 39; tripartite motif-containing 39; Tripartite motif-containing protein 39
Gene Aliases: 1100001D15Rik; E130103K13Rik; mKIAA4179; RBCC-B30.2; RNF23; TFP; TRIM39; TRIM39B
UniProt ID: (Human) Q9HCM9, (Rat) Q6MFZ5, (Mouse) Q9ESN2
Entrez Gene ID: (Human) 56658, (Rat) 309591, (Mouse) 79263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support