Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
TRIM5a is a 493 amino acid member of the large tripartite motif protein (TRIM) family. TRIM proteins are composed of three zinc-binding domains, a RING, a B-box 2 and a coiled-coil domain, and they use homomultimerization to identify different cell compartments. Some TRIM proteins, such as TRIM5a, also possess a carboxy-terminal B30.2 (SPRY) domain and localize to the cytoplasm. TRIM5a mediates innate intracellular retroviral resistance, which is dependent on its carboxy-terminal domain. The three variable regions of the B30.2 domain form loops on one side of the B30.2 core structure of TRIM5a which may form a binding surface for the virus. TRIM5a trimerization plays a major role in its affinity for the retroviral capsid, and in its ability to inhibit virus infection. The linker region between the coiled-coil and B30.2 domains of TRIM5a is required for this trimerization. TRIM5a blocks infection after the virus has entered the cell.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RING finger protein 88; RING-type E3 ubiquitin transferase TRIM5; trim5a; tripartite motif containing 5 transcript variant iota; tripartite motif containing 5 transcript variant kappa; tripartite motif protein TRIM5; Tripartite motif-containing protein 5
Gene Aliases: EG667823; Gm8833; RNF88; TRIM5; TRIM5alpha
UniProt ID: (Human) Q9C034
Entrez Gene ID: (Human) 85363, (Mouse) 667823
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support