Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MEEELAIQQ GQLETTLKEL QTLRNMQKEA IAAHKENKLH LQQHVSMEFL KLHQFLHSKE KDILTELREE GKALNEEMEL NLSQLQEQCL LAKDMLVSIQ AKTEQQNSFD FLKDITTLLH SLEQGMKVLA TRELISRKLN LGQYKGPIQY MVWREMQDTL CPGLSPLTLD PKTAHPNLVL SKSQTSVWHG DIKKIMPDDP ERFDSSVAVL GSRGFTSGKW YWEVEVAKKT KWTVGVVRES IIRKGSCPLT PEQGFWLLRL RNQTDLKALD LPSFSLTLTN NLDKVGIYLD YEGGQLSFYN AKTMTHIYTF SNTFMEKLYP YFCPCLNDGG ENKEPLHILH PQ (1-341 aa encoded by BC033314)
This gene encodes a member of the RING-B-box-coiled-coil (RBCC) family and encodes a protein with an N-terminal RING finger motif, a PRY domain and a C-terminal SPRY domain. The mouse ortholog of this gene is specifically expressed in germ cells at the round spermatid stages during spermatogenesis and, when overexpressed, induces apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM69; hCG_39321; RFP-like (B30.2) domain protein; RFP-like domain-containing protein trimless; RING finger B-box coiled-coil transcription factor; RING finger protein 36; RING-type E3 ubiquitin transferase TRIM69; Testis-specific RING finger protein; Tripartite motif-containing protein 69
Gene Aliases: 4921519C19Rik; HSD-34; HSD34; RNF36; Trif; TRIM69; Trimless
UniProt ID: (Human) Q86WT6, (Rat) Q5BK82, (Mouse) Q80X56
Entrez Gene ID: (Human) 140691, (Rat) 311373, (Mouse) 70928
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support